Analytical guard column, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-130B
UPCHC-130BEA 378 EUR InStock
UPCHC-130B
Analytical guard column, Upchurch Scientific®
Chromatographiesäulen
This narrow-bore short column (2,0×20 mm) creates only a slight pressure increase with virtually no detectable theoretical plate loss when used with a 3 mm int.Ø or larger column.
  • Available as a kit with funnel and extra frits
  • Easy to pack
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge