Bosshead (sleeve)
Lieferant: USBECK Laborgeräte
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2240
USBE2240EA 6.6 EUR InStock
USBE2240
Bosshead (sleeve)
Klemmen Stativmuffen
Bosshead made of powder-coated zinc die-cast, with nickel plated brass screws.
  • For rods up to 16 mm Ø
  • Location hole 10 mm Ø
Weight: 80 g
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge