Anti-CTGF Mouse Monoclonal Antibody [clone: 558CT34.4.4]
Lieferant: US Biological
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
030895
USBI030895EA 667 EUR InStock
USBI030895
Anti-CTGF Mouse Monoclonal Antibody [clone: 558CT34.4.4]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100
Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Monoclonal
Clone: 558CT34.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge