Anti-UNC13D Goat Polyclonal Antibody
Lieferant: US Biological
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M2196-85G
USBIM2196-85GEA 632 EUR InStock
USBIM2196-85G
Anti-UNC13D Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: UNC13D (unc-13 homolog D (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge