Anti-NuMA Mouse Monoclonal Antibody [clone: A73-B/D12]
Lieferant: US Biological
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N8341-05
USBIN8341-05EA 843 EUR InStock
USBIN8341-05
Anti-NuMA Mouse Monoclonal Antibody [clone: A73-B/D12]
Antikörper
Type: Primary
Antigen: NuMA
Clonality: monoclonal
Clone: A73-B/D12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge