Human Recombinant tPA (From CHO cells)
Lieferant: UNITED BIOCHEMICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
T5600-07B
USBIT5600-07BEA 776 EUR InStock
USBIT5600-07B
Human Recombinant tPA (From CHO cells)
Proteine und Peptide
This recombinant human tPA is active and expressed in Chinese hamster ovary (Cho) cells. Fully complexable with PAI-1.
  • Activity: >90% active as determined by titration with PAI-1
Achtung! Bitte beachten: Intended for research use only. Not for use in human, therapeutic, or diagnostic applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge