Refill earplugs for "one 2 click" dispenser
Lieferant: UVEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2112.003 2112.023 2112.118 2112.022
UVEX2112.003EA 64.6 EUR InStock
UVEX2112.003 UVEX2112.023 UVEX2112.118 UVEX2112.022
Refill earplugs for "one 2 click" dispenser
Gehörschutzstöpsel Gehörschutzstöpsel, zur einmaligen Verwendung
Refill packs and boxes for the uvex x-fit earplug dispenser "one 2 click".
uvex x-fit: The ergonomically shaped uvex x-fit disposable earplug is characterised by very high sound absorption levels, making it suitable for use in very loud environments. The uvex x-fit foam earplug has a sealed surface, preventing foreign bodies from penetrating. The soft foam offers real comfort, even when worn for extended periods. Ideal for use in very loud environments. SNR = 37 dB.
uvex com4-fit: With its compact, ergonomically shaped design, the uvex com4-fit is comfortable even for wearers with narrower ear canals. The uvex com4-fit has a sealed surface, reducing the risk of contamination penetrating the plugs. The soft foam provides a high level of comfort, even when worn for long periods. SNR = 33 dB.
uvex hi-com: The innovative shape of the uvex hi-com earplugs reduces the level of sound that normally builds up in the inner ear when wearing hearing protection. Integrated cavities in the direction of the eardrum result in a larger surface area which absorbs more noise The earplugs provide softer acoustics and absorb disruptive noise, as well as allowing excellent speech recognition. SNR = 26 dB.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge