Safety shoes, lace-up, uvex suXXeed®, 62553
Lieferant: UVEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6255344 6255345 6255336 6255337 6255346 6255347 6255338 6255339 6255340 6255335 6255341 6255342 6255343
UVEX6255344EA 76.4 EUR InStock
UVEX6255344 UVEX6255345 UVEX6255336 UVEX6255337 UVEX6255346 UVEX6255347 UVEX6255338 UVEX6255339 UVEX6255340 UVEX6255335 UVEX6255341 UVEX6255342 UVEX6255343
Safety shoes, lace-up, uvex suXXeed®, 62553
Schuhe und Stiefel
Metal-free, flexible, lightweight safety shoe made from oiled hydro suede leather upper, resistant to liquid penetration, collar and gusset in water-repellent fabric, fabric eyelets plus a double-desity PU2D sole , highly slip resistant and resistant to oils and hydrocarbons.
  • Quick drying Climate Airmesh lining, very resistant to abrasion, very good abrasion resistance and very good water vapour absorption
  • Removable insole, top in abrasion resistant fabric
  • Metal-free protective toecap (200 J)
  • Metal-free puncture resistant midsole
Suitable for heavy industry, warehousing and logistics.
Colour: Black/red
Zertifizierungen: EN ISO 20345:2011; S3, SRC
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge