ESD safety shoes, uvex motion style, 6978
Lieferant: UVEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6978.838 6978.839 6978.836 6978.837 6978.840 6978.841 6978.842 6978.843 6978.844 6978.845 6978.846 6978.847 6978.848
UVEX6978.838EA 137 EUR InStock
UVEX6978.838 UVEX6978.839 UVEX6978.836 UVEX6978.837 UVEX6978.840 UVEX6978.841 UVEX6978.842 UVEX6978.843 UVEX6978.844 UVEX6978.845 UVEX6978.846 UVEX6978.847 UVEX6978.848
ESD safety shoes, uvex motion style, 6978
Schuhe und Stiefel
Ultra-light ESD safety shoe made from breathable high-tech micro velour, soft padded collar and tongue, breathable distance mesh plus an extremely flexible dual-density PUR sole (slip, abrasion and cut resistant, resistant to oil and petrol also withstands brief exposure to heat of approx. +120 °C). Free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/grey
Width: 11
Zertifizierungen: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S2, SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge