Ultrasonic Planet Goggles
Lieferant: UVEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9302290
UVEX9302290EA 32.1 EUR InStock
UVEX9302290
Ultrasonic Planet Goggles
Vollsichtbrillen
The combination of hard and soft material components used in the uvex ultrasonic pairs high mechanical strength with optimum wearability.
  • Functional goggles with panoramic lens for a wide range of applications
  • Metal free PC lens
  • Headband: anthracite, jade
  • Soft, flexible face pad naturally moulds to many facial shapes
  • Indirect ventilation system for increased air circulation
  • Length-adjustable fabric headband for a secure fit and individual adjustment
  • UV400 – 100% protection against harmful UV radiation to 400 nm
  • Increased mechanical strength – certification B (120 m/s)
This wide-vision goggle can be worn for long periods without causing any pressure problems. The flexible soft component allows it to adapt to each individual face for a snug fit, ensuring comprehensive side protection around the eye area, even when worn over prescription spectacles. The uvex ultrasonic offers outstanding ventilation, ensuring a pleasant interior environment that does not cause irritation.
uvex protecting planet – mass-balanced PC lens, frame, headband and packaging partially made from recycled material. Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens. Generous internal volume to accommodate prescription spectacles and ensure a comfortable fit without pressure points.
Zertifizierungen: Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
Certification: W 166 34 B CE – 2C-1.2 W1 BKN CE
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge