Chemistry diaphragm pump, MV 10C NT
Lieferant: VACUUBRAND
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20744300
VACU20744300EA 8460 EUR InStock
VACU20744300
Chemistry diaphragm pump, MV 10C NT
Pumpen Vakuum-Membranpumpen
This four-stage chemistry-design diaphragm pump is an excellent solution for continuous, oil-free pumping of corrosive gases and vapors and meet highest requirements. The four-stage design of the eight-cylinder pump MV 10C NT provides the advantageous combination of high pumping speed and very low ultimate vacuum of 0,9 mbar in a very compact design. All internal parts in contact with pumped media are made of chemically resistant fluoroplastics. Well-proven PTFE sandwich diaphragms increase reliability and extend operating life.
  • Outstanding chemical resistance and superior vapor tolerance
  • Reduced process time due to very high flow rates even close to ultimate vacuum
  • Very low vibration and quiet
  • Excellent ultimate vacuum even with gas ballast
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge