Rotary Vane Pumps, RE 2.5
Lieferant: VACUUBRAND
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20797150
VACU20797150EA 1725 EUR InStock
VACU20797150
Rotary Vane Pumps, RE 2.5
Pumpen Vakuum-Drehschieberpumpen
Rotary vane pumps from VACUUBRAND offer great performance in a compact design. Even with the gas ballast open, they enable a very good ultimate vacuum. You also benefit from extended oil change and maintenance intervals thanks to optimised lubrication circuit and large usable oil volume.
  • High flow rates even at vacuum levels approaching ultimate vacuum
  • Vacuum-tight at switch-off; external anti-suckback valve not needed
  • Large oil volume: Long intervals between oil changes
  • Ease of maintenance due to telescopic design
The one-stage pumps RE 2,5 are high-performance rotary vane pumps with extra compact design and low weight. They are the ideal solution for a wide range of laboratory and process applications that require low ultimate vacuum at medium gas flow rate. Typical applications are lyophilisation, fine vacuum distillation and evacuation of small volumes, e.g., for utilisation of µ-focus x-ray tubes or filling with pure gases.
Lieferumfang: Pump oil filled and completely mounted, ready for use, with manual.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge