Rotary Vane Pumps, RZ 9
Lieferant: VACUUBRAND
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20798140
VACU20798140EA 3950 EUR InStock
VACU20798140
Rotary Vane Pumps, RZ 9
Pumpen Vakuum-Drehschieberpumpen
Rotary vane pumps from VACUUBRAND offer great performance in a compact design. Even with the gas ballast open, they enable a very good ultimate vacuum. You also benefit from extended oil change and maintenance intervals thanks to optimized lubrication circuit and large usable oil volume.
  • Very high flow rates even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance due to efficient gas ballast
  • Very good ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off
  • External anti-suckback valve not needed
  • Large oil volume: long intervals between oil changes
  • Ease of maintenance due to telescopic design
The RZ 9 is a two-stage powerful rotary vane pump for fast evacuation of vessels and at higher gas flow rates. Typical applications are lyophilization, fine vacuum distillation and drying chambers. The two-stage rotary vane pump RZ 9 is also available in various packages with matching accessories such as exhaust filter FO and valve VS 25. Rotary vane pumps are used for a process vacuum of a few mbar to 10⁻³ mbar. 40 mbar water vapor tolerance.
Lieferumfang: Pump oil filled and completely mounted, ready for use, with manual.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge