Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Chemistry-HYBRID pump, RC 6

Lieferant: VACUUBRAND

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

20798560
VACU20798560EA 5070 EUR InStock
VACU20798560
Chemistry-HYBRID pump, RC 6
Pumpen Vakuum-Drehschieberpumpen
The RC 6 chemistry-HYBRID pump combines a two-stage rotary vane pump and a two-stage chemistry diaphragm pump for optimized corrosion resistance.

  • Reduced internal corrosion, even when working with corrosive vapours
  • Oil changes typically reduced 90% or more compared with rotary vane pumps alone
  • Most economical solution: In practical operation a cold trap is often no longer necessary
  • Ease of maintenance due to telescopic design

The RC 6 chemistry-HYBRID pump is a combination of a two-stage rotary vane pump and a two-stage chemistry diaphragm pump for optimized corrosion resistance. The diaphragm pump maintains the oil reservoir under vacuum in order to keep the partial pressures of solvent vapours at levels below their condensation points and to reduce largely the concentration of oxygen and corrosive gases. Therefore, the RC 6 chemistry-HYBRID pump has a much higher solvent vapour pumping capability and resistance to aggressive gases than conventional rotary vane pumps.

Water vapor tolerance with gas ballast >40 mbar.

Lieferumfang: Pump completely mounted, ready for use after oil filling (bottle 0,5 L enclosed), with manual.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge