Solid phase extraction cartridges, Bond Elut™ Sodium Sulphate Drying
Lieferant: VARIAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
12162051B 12162052B 12162054B 12131033 12132004
VARI12162051BEA 386 EUR InStock
VARI12162051B VARI12162052B VARI12162054B VARI12131033 VARI12132004
Solid phase extraction cartridges, Bond Elut™ Sodium Sulphate Drying
Chromatographiesäulen
Bond Elut™ Jr cartridges have top and bottom Luer fittings allowing easy sample processing when used in conjunction with standard SPE cartridges. Bond Elut™ LRC cartridges have a large reservoir above the sorbent bed and are suitable for use on any standard SPE vacuum manifold.
  • Highly effective pre-packed dessicant
  • Clean ACS grade, anhydrous sodium sulphate
  • Prepacked for convenience
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge