Deactivated injector liners
Lieferant: VARIAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CP14988
VARICP14988EA 1370 EUR InStock
VARICP14988
Deactivated injector liners
Chromatographie-Zubehör Chromatographie-Liner
These deactivated injector liners are optimised for sensitive and reproducible analysis.
  • Easy-opening storage trays
  • High performance standards
  • Silanisation activation prevents sample degradation
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge