RheBuild® kits for injection valves
Lieferant: VARIAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RE7125079 RE7725999
VARIRE7125079EA 0 EUR InStock
VARIRE7125079 VARIRE7725999
RheBuild® kits for injection valves
Chromatographie-Zubehör Chromatographie-Ersatzteile
RheBuild® kits containing genuine Rheodyne® parts are available for most Rheodyne® products. RheBuild® kits eliminate the need to order individual parts.
Lieferumfang: Each RheBuild® kit includes all the parts, tools and instructions to maintain the precision performance of Rheodyne® valve platforms.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge