Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Conjugation kits, protein-oligo conjugation kit

Lieferant: VECTOR LABORATORIES

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


S-9011-1
VECTS-9011-1EA 1350 EUR InStock
VECTS-9011-1
Conjugation kits, protein-oligo conjugation kit
Tests Proteintests
The Protein-Oligo Conjugation kit is designed to easily and efficiently conjugate two separate protein-oligo conjugates. This kit is flexible so that researchers with little or no conjugation experience can make their own custom protein-oligo conjugates to suit their needs. It includes all of the necessary components - including S-HyNic and S-4FB - and protocols for easy and specific crosslinking of any protein with any oligo up to 100 base pairs in length.

  • Catalysed conjugation – Faster kinetics for greater efficiency and yields
  • Quantifiable – Using a UV signature wavelength and simple UV scan
  • Stable – 10 times more stable than any other conjugation linker
  • Specificity – Two-linker method avoids homoconjugate formation


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge