Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

High volume magnetic stirrers

Lieferant: VELP SCIENTIFIC

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

F203A0511 F203A0513 F203A0510 F203A0512
VELPF203A0511EA 570 EUR InStock
VELPF203A0511 VELPF203A0513 VELPF203A0510 VELPF203A0512
High volume magnetic stirrers
Rührer Magnetrührer
Magnetic stirrers that guarantee continuous operation at both low and high speeds ensuring safety and resistance to chemical agents.

  • The brushless motor provides continuous operation with high liquid volumes at any speed
  • Premium power and long life span
  • Constant stirring from 40 to 1500 rpm (MSL 8 - 25) or from 20 to 1100 (MSL 50) with SpeedServo technology
  • Gentle start and low speed to protect samples
  • The control panel is protected from liquid spills
  • Resistant stainless steel plate are
  • Designed for a long lifespan and guaranteed with 2 years warranty
  • Timer, auto-reverse, intermittent mode

Due to the brushless motor, the stirrers stay cold even after several days of continuous operation. This feature makes it particularly suitable for use in microbiology and biochemistry. Epoxy painted metal exterior, with stainless steel plate, stirrers have been designed and tested to give excellent resistance to chemical agents and corrosion in general. The stirring speed can be set within a range of 40 to 1500 rpm using the knob on the front panel.

Zertifizierungen: EN 61010-1, EN 61010-2-051, EN 61326-1, 2006/42/EC, 2014/30/EU
2011/65/UE (RoHS), 2012/19/UE (RAEE)
CB Certificate
IEC 61010-1:2010, IEC 61010-1:2010/AMD1:2016
IEC 61010-2-051:2015
cNEMKOus
UL std. No 61010-1-2nd

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge