Nitrogen analyser
Lieferant: VELP SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F30800080
VELPF30800080EA 0 EUR InStock
VELPF30800080
Nitrogen analyser
Analysatoren Protein-Analysatoren
Powerful protein/nitrogen analyser able to perform precise analysis and determination in a flash for high productivity and unique benefits.
  • Controllable from PC with DUMAsoft™ software for detailed reporting
  • Automated analysis of preloaded samples for over 100 positions
  • User-selectable methods with storable data
Zertifizierungen: Conforms to Good Laboratory Practice standard.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge