Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

AM 4 Digital Multi-position Hotplate Stirrers

Lieferant: VELP SCIENTIFIC

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

F20500600 SA20500600
VELPF20500600EA 2260 EUR InStock
VELPF20500600 VELPSA20500600
AM 4 Digital Multi-position Hotplate Stirrers
Rührer Mehrstellen-Magnetrührer mit Heizung
Digital hotplate stirrers with four independent plates in CerAlTop™, ensuring precise thermoregulation of the medium, high safety, and maximum reliability. Units have epoxy painted aluminum housing.

  • Four independent positions which stir from 50 to 1500 rpm, up to 20 L each
  • The 135 mm diameter CerAlTop™ plates ensure optimum temperature homogeneity and superior resistance from chemicals and scratches
  • Precise thermoregulation of the medium with PT100/PT1000 probe (±1.0 °C) and VTF Digital Thermoregulator (±0.5 °C)
  • Hot surface safety warning displayed when the temperature is above 50 °C
  • Brushless motor ensuring power and resistance over time
  • Designed for long lifespan and guaranteed with 3 years warranty with online product registration

The AM 4 Digital and the Digital System has external sensor connections for PT100; PT1000 and VTF probes. Units have an epoxy painted aluminum housing.

Zertifizierungen: CB; cNEMKOus

Lieferumfang: The AM 4 Digital - System is supplied with AM 4 Digital, support rod (×4), PT100 Probe AISI 316 Ti Ø3 mm (×4), clamp for probe (×4).

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge