Tygon S3™ B-44-3 Tubing
Lieferant: Saint-Gobain Performance plastics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AAB00046
VERNAAB00046EA 1040 EUR InStock
VERNAAB00046
Tygon S3™ B-44-3 Tubing
Schläuche Transferschlauch
Tygon S3™ B-44-3 tubing for beverage transfer from Saint-Gobain is phthalate-free and specifically formulated to ensure it does not affect the taste nor odour of liquids moving through it, including soft drinks and juices.
  • Clear as glass for easy visual monitoring of flow
  • Lightweight and flexible for easy, quick installation
  • Broad chemical resistance
  • Non-wetting properties allow easy cleaning and complete drainage
  • Contains no BPA or phthalates
  • Autoclavable
It is non-wetting, which allows it to completely drain and flush clean with ease, and its glass-like clarity makes it easy to monitor and adjust the beverage flow during processing. Light in weight and easy to handle, Tygon S3 B-44-3 tubing can be put into service quickly. It readily bends to accommodate abrupt corners and obstructions, requiring a minimium of couplings and fittings. It is ideal for applications such as food and beverage transfer and dispensing.
Hardness: 63 (shore A)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge