Tube clamps, Pure-Fit® TCL
Lieferant: Saint-Gobain Performance plastics
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PFTCL150 PFTCL250
VERNPFTCL150EA 2270 EUR InStock
VERNPFTCL150 VERNPFTCL250
Tube clamps, Pure-Fit® TCL
Klemmen Schlauchklemmen und Sanitärklemmen
Pure-Fit® TCL large tube clamps provide the same innovative clamping technology as Pure-Fit TC but are designed to accommodate large diameter and thick wall tubing applications.
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Custom colours and identification available
  • Fully autoclavable and sterilisable
Pure-Fit TCL is compatible with standard, braided (reinforced) and multilayer tubing. In addition to providing complete fluid stoppage, Pure-Fit TCL can be used in the flow position and features an integral bore hole for absolute lock-out capabilities.
Pure-Fit® TCL is constructed from animal derivative-free PVDF. Pure-Fit TCL is available in two sizes to accommodate a wide range of thick wall tubing and can be used with up to 2,0" O.D. tubing.
Zertifizierungen: Meets USP Class VI criteria.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge