Gas syringes, series A-2, Pressure-Lok®
Lieferant: VICI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PS-050025 PS-050031 PS-050032 PS-050034 PS-050035
VICIPS-050025EA 194 EUR InStock
VICIPS-050025 VICIPS-050031 VICIPS-050032 VICIPS-050034 VICIPS-050035
Gas syringes, series A-2, Pressure-Lok®
Spritzen Spritzen für die Chromatographie
The syringes feature a PTFE plunger tip, stress-formed by a special process to ensure a leak-tight seal. The self-lubricating plunger tip stays smooth for the life of the syringe, with none of the seizing or residue buildup associated with conventional all-metal plungers.
  • Removable needles
  • PTFE plunger tip
  • PTFE-sealed needle
  • Low dead volume
  • Crisp clean graduations
Small liquid samples with low-boiling components are not lost through evaporation, as often occurs with ordinary syringes. The positive rear stop prevents plunger blowout at elevated pressures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge