Cleanroom wipes, PolyControl
Lieferant: FHP VILEDA SCS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
142872_U
VILE142872_UEA 64.8 EUR InStock
VILE142872_U
Cleanroom wipes, PolyControl
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
These disposable wipes are made of 100% Vileda Professional polyester fibre. They are ideal for standard cleanroom cleaning applications.
  • Optimum stitch density for specific applications
  • Cleanroom laundered and packed
  • Sealed edges via laser cutting for low particle release
PolyControl 400: for ISO class 4 - 9 environments
- Rough surface wipe
- Ultra low level of particles and extractables
- Resists abrasion under rigorous use
PolyControl 600: for ISO class 5 - 9 environments
- Delicate surface wipes
- Low level of particles and extractables
- Fine polyester fibres with high absorbency
PolyControl 900: for ISO class 4 - 9 environments
- High absorbency and spill pick-up wipes
- Ultra low level of particles and extractables
- Unique two-ply construction for excellent absorbency
Colour: White
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge