Pipette controller, VITLAB pipeo®
Lieferant: VITLAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1631500
VITL1631500EA 452 EUR InStock
VITL1631500
Pipette controller, VITLAB pipeo®
Pipetten Pipettierhelfer
This pipette controller holds pipettes from 0,1 to 200 ml securely and tightly in the exchangeable adapter. Liquid vapours are purged directly to protect the instrument.
  • Ergonomic handle – very lightweight at about 190 grams – and excellent balance
  • Speed can be adjusted easily, continuously and exactly with one hand using two buttons
  • 50 ml pipette can be filled comfortably in less than ten seconds
  • Liquid release can be done either by gravity delivery when calibrated 'Ex' (to deliver), or in blow out mode using the battery-operated motor
  • One full charge of the nickel-metal hydride battery allows 8 hours of non-stop pipetting
  • Charge level of the recyclable battery is shown by the LED indicator
  • Pipette controller can be used while the battery is being recharged
Lieferumfang: Supplied with battery charger (100 to 240 V, 50/60 Hz), four plug adapters (EU, UK, US/J, AUS), battery, battery compartment cover, two replacement 0,2 µm membrane filters and instruction manual.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge