Wash bottles, narrow or wide neck
Lieferant: VITLAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
94993 93793 95093 93993 95193 94193
VITL94993EA 5.4 EUR InStock
VITL94993 VITL93793 VITL95093 VITL93993 VITL95193 VITL94193
Wash bottles, narrow or wide neck
Flaschen Spritzflaschen
PP, translucent, screw cap and spray tube made from PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge