Filter for ultrapure water system, arium® CellPlus
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H2O-CUF
VIVAH2O-CUFEA 314 EUR InStock
VIVAH2O-CUF
Filter for ultrapure water system, arium® CellPlus
Filter Flüssigkeits-Prozessfilter
The arium® CellPlus is a sterile packed, point-of-use ultrafilter for efficient removal of endotoxins, RNase, DNase, microorganisms and particles. Retrograde contamination is prevented by the protective bell design made up of polycarbonate. It offers high safety for critical cell culture applications.
  • Effective removal of RNase and DNase
  • Reliable removal of endotoxins
  • High flow rate performance
  • Certified quality
The high grade material enables excellent total throughputs and optimal flow rates. Designed for arium® comfort and arium® mini ultrapure water systems.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge