Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

VWR® Square Polycarbonate Media Bottles

Lieferant: VWR Collection

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

VWRI210-000094EA 181 EUR InStock
VWRI210-000094 VWRI210-000102 VWRI210-000095 VWRI210-000103 VWRI210-000096 VWRI210-000098 VWRI210-000087 VWRI210-000089 VWRI210-000086 VWRI210-000091 VWRI210-000088 VWRI210-000085
VWR® Square Polycarbonate Media Bottles
Flaschen Kunststoffflaschen
VWR® square polycarbonate bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −130 to +121 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.

  • Made from durable 100% virgin polycarbonate material
  • Suitable for frozen storage down to −130 °C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and closures (HDPE/PP) provided for applicable products
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability

Bottles are arranged in shrink-wrapped trays and polybag to prevent contamination. 2 L bottles feature a moulded hand grip for better ergonomics. Bottles 30 ml and above have volume graduations.

Zertifizierungen: US FDA 21CRF, USP CLASS VI compliant Resin.

Zubehörinformationen: Spare closures are available for specific application requirements. See part numbers VWRI210-000111, VWRI210-000112, VWRI210-000113, VWRI210-000114, VWRI210-000115, VWRI210-000116, VWRI210-000117, VWRI210-000118, VWRI210-000119 and VWRI210-000120.

Verpackung: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge