VWR® Centrifuge Tube Rack, 15 and 50 ml
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VWRI479-2289EA 693 EUR InStock
VWRI479-2289
VWR® Centrifuge Tube Rack, 15 and 50 ml
Gestelle Reagenzglasgestelle
Racks are specially designed to provide an ideal storage solution for large quantities of samples in 50 ml centrifuge tubes.
  • Easy access and storage
  • Security lock devices are available in each rack
  • Made of corrosion resistant pharmaceutical grade stainless steel
Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge