Accessories for Endotoxin Tests
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
293-51601
WAKO293-51601EA 0 EUR InStock
WAKO293-51601
Accessories for Endotoxin Tests
Mikrobiologische Testsysteme Endotoxin-Tests
This endotoxin-free solution containing human serum albumin (HSA) is capable of extracting endotoxins that cannot be extracted in water or saline solution.
Traditionally, water or saline solution has been used to extract endotoxin in tests involving medical devices and equipment; however, the efficacy of this extraction method is debated. By using this extraction solution, it provides our customers with a more reliable method for medical device testing.
Recommended for use in endotoxin testing on equipment and devices that may come into contact with fluids containing blood or protein.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge