Copolyester BPA-Free Blender Container With Blade, CAC139
Lieferant: WARING BLENDOR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CAC139
WARICAC139EA 402 EUR InStock
WARICAC139
Copolyester BPA-Free Blender Container With Blade, CAC139
Homogenisatoren Standard-Labormischer
The CAC139 is the clear copolyester 1,36 L blender container from Waring with the blade assembly included. These containers are designed for the TORQ 2.0 TBB series blenders.
  • Durable copolyester construction
  • Blade assembly included
  • Copolyester material
  • Clear colour
These blender containers are dishwasher-safe, stackable, and BPA-free.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge