Surfactant, RapiGest™ SF
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
186001861
WATE186001861EA 375 EUR InStock
WATE186001861
Surfactant, RapiGest™ SF
Protein-Extraktion Proteinextraktions-Reagenzien
RapiGest™ SF is an anionic surfactant for peptide digestion. Supplied as five 1 mg vials for 1 to 50 digestions. The digestion speed and more peptide recovery is enhanced by using trypsin or other enzymes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge