UPLC columns, ACQUITY UPLC™ BEH phenyl
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
186002884 186002885 186003375 186003911
WATE186002884EA 1930 EUR InStock
WATE186002884 WATE186002885 WATE186003375 WATE186003911
UPLC columns, ACQUITY UPLC™ BEH phenyl
Chromatographiesäulen
These columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Chemically stable phenyl sorbent
  • Ultra-low column bleed, long column lifetimes and excellent peak shape
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge