Columns, ACQUITY UPLC™ HSS T3
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
186003538
WATE186003538EA 2130 EUR InStock
WATE186003538
Columns, ACQUITY UPLC™ HSS T3
Chromatographiesäulen
An universal, silica based bonded phase with ACQUITY HSS T3 sorbents which is used for the separations for polar and non-polar compounds. It is compatibility with 100% of aqueous mobile phase. It exhibit ultra-low MS bleed, scalable and transferable to HPLC separations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge