Gel permeation chromatography columns, Styragel® HR
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WAT044231 WAT054405 WAT054415 WAT044229 WAT044228 WAT044237 WAT044222
WATEWAT044231EA 4370 EUR InStock
WATEWAT044231 WATEWAT054405 WATEWAT054415 WATEWAT044229 WATEWAT044228 WATEWAT044237 WATEWAT044222
Gel permeation chromatography columns, Styragel® HR
Chromatographiesäulen
Styragel® HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterising samples in the low to mid molecular range where high resolution is critical.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge