Qualitative filter papers, standard grades, grade 597 Optima, Whatman™
Lieferant: GE Healthcare - Whatman
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
989410108 989410109 989410107 989410106 989410105 989410110 98949613 989410104 989410103 98949329 989410102 98949552 989410101
WHAT989410108EA 62.2 EUR InStock
WHAT989410108 WHAT989410109 WHAT989410107 WHAT989410106 WHAT989410105 WHAT989410110 WHAT98949613 WHAT989410104 WHAT989410103 WHAT98949329 WHAT989410102 WHAT98949552 WHAT989410101
Qualitative filter papers, standard grades, grade 597 Optima, Whatman™
Filter Filterpapiere Qualitatives Filterpapier
Standard Whatman grade filter paper for a variety of routine applications in food and beverage industries. Well-suited for determination of fat content in food or removal of carbon dioxide and turbidity from beverages such as beer.
  • Available in sheets and circles
  • Medium fast filtration rate
  • Minimum 90% cellulose content
  • Technical and scientific support
These cellulose filters are used in qualitative analytical techniques to determine and identify materials.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge