Arsine generator apparatus, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
180034 180030 970440
WHEA180034EA 180 EUR InStock
WHEA180034 WHEA180030 WHEA970440
Arsine generator apparatus, WHEATON®
Aufschlusssysteme Geräte zum Nachweis von Spurenelementen
Designed for arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method.
  • Assembled apparatus is freestanding and easy to use
  • Design eliminates the ball and socket joint connecting the scrubber and absorber components
Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge