Media bottles, safety-coated, without screw cap, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
219457 219455
WHEA219457EA 1050 EUR InStock
WHEA219457 WHEA219455
Media bottles, safety-coated, without screw cap, WHEATON®
Flaschen Zellkulturflaschen
USP Type I clear borosilicate glass, Plastisol coated.
  • Provided with graduations and writing patch
  • 33-430 screw cap can be purchased separately
  • Autoclavable
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge