Plier decappers, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
224372 224373
WHEA224372EA 256 EUR InStock
WHEA224372 WHEA224373
Plier decappers, WHEATON®
Crimper, Capper und Decapper
These pliers decappers are constructed of steel with grey plastic coated handles. To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge