Headspace vials, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
225277 225280 W225282
WHEA225277EA 98.5 EUR InStock
WHEA225277 WHEA225280 WHEAW225282
Headspace vials, WHEATON®
Vials
These vials made of type I borosilicate glass feature rounded shoulders and bottoms, allowing for even heating and safer operation at higher pressures.
  • Accepts 20 mm aluminium seal
  • Top designed to provide a tight seal with septa
Bestellinformation: Aluminium seals must be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge