Caps for bottles, open top, Microlink®, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
240106 240235-01 240250 240241 240241-01 240250-01 240251-01 240256-01 240112 240114 240200
WHEA240106EA 143 EUR InStock
WHEA240106 WHEA240235-01 WHEA240250 WHEA240241 WHEA240241-01 WHEA240250-01 WHEA240251-01 WHEA240256-01 WHEA240112 WHEA240114 WHEA240200
Caps for bottles, open top, Microlink®, WHEATON®
Verschlüsse Flaschenverschlüsse
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
  • PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
  • Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge