Trypsinising flasks, with screw caps, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
355394 355395 355397 355392 355445 355442 355444
WHEA355394EA 145 EUR InStock
WHEA355394 WHEA355395 WHEA355397 WHEA355392 WHEA355445 WHEA355442 WHEA355444
Trypsinising flasks, with screw caps, WHEATON®
Kolben Trypsinisierungskolben
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Zertifizierungen: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge