Vacuum filtration complete units, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
419347
WHEA419347EA 462 EUR InStock
WHEA419347
Vacuum filtration complete units, WHEATON®
Trichter Membranfiltertrichter
Vacuum filter holder with fritted glass support base complete.
  • Recommended for general filtration
  • Flasks (not included) have a hose connection for ¼" (6 mm) Int.Ø tubing
  • Very good chemical and temperature resistance
Designed to handle up to 1000 ml of liquid for analysis of particulate or microbiological contamination.
Glass components manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Lieferumfang: Includes a coarse porosity (40 to 60 µm) fritted glass support base, 300 ml graduated funnel, anodized aluminum clamp, and a No. 8 silicone stopper. Flask not included.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge