WHEATON® Bottle Top Vacuum Filtration System
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
419520-47 419505 419530-47
WHEA419520-47EA 936 EUR InStock
WHEA419520-47 WHEA419505 WHEA419530-47
WHEATON® Bottle Top Vacuum Filtration System
Filter Flaschenaufsatzfilter
The system is designed to collect the filtrate directly into a standard laboratory bottle.
Glass components are manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements. Connection to the bottle is made with a polybutylene terephthalate (PBT) cap and a PTFE-faced silicone sealing ring.
Lieferumfang: Includes a coarse porosity (40 to 60 µm) fritted glass support base, graduated funnel (300 ml), anodised aluminium clamp, and reservoir (1 L).
Order Now
PRODUKT
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge