Pipette tips, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
851358
WHEA851358EA 175 EUR InStock
WHEA851358
Pipette tips, WHEATON®
Pipettenspitzen
These high quality tips are made of high grade, virgin, metal-free PP.
  • Compatible with most macro pipettes
  • Precision injection molding care for air-tight fit
  • Free from trace metals for excellent accuracy and reproducibility
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge