Sample bottles, narrrow neck, with screw cap, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
990477
WHEA990477EA 108 EUR InStock
WHEA990477
Sample bottles, narrrow neck, with screw cap, WHEATON®
Flaschen Glasflaschen
Glass, clear.
  • Excellent for accessing fluid flow in deep manholes and hard to reach sub-surface sampling areas
  • Helps meet EPA requirements for water and waste water sampling
  • 38-430 thread
Each sampler consists of an inert polypropylene head with stainless steel fittings and an aluminum pole assembly with a golden alodine finish. It comes with a 1000 mL narrow-mouth sample bottle with a PTFE- lined cap. To use it, screw a sample bottle into the head and lower to the desired depth. Pull the cable at the top of the handle which will lift a spring-loaded plunger from the bottle opening, allowing liquid to enter through four 5/8" diameter holes. Release the cable when the bottle is full to reseat the plunger.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge