Rubber stoppers for lyophilisation, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
W224100-282 224100-203 W224100-193 W224100-093 W224100-202
WHEAW224100-282EA 1390 EUR InStock
WHEAW224100-282 WHEA224100-203 WHEAW224100-193 WHEAW224100-093 WHEAW224100-202
Rubber stoppers for lyophilisation, WHEATON®
Stopfen Gummistopfen
Ultra pure chlorobutyl-isoprene rubber stoppers for lyophilisation.
  • Very low level of moisture adsorption and absorption of the preservatives in aqueous solution
  • Reduced possibility of the legs sticking to each other
  • Hardness of the lyophilization stopper is 46 Shore A
Designed for use with vials and serum bottles.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge