Lyophilisationsampullen, Vacule®, WHEATON®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
W651446
WHEAW651446EA 919 EUR InStock
WHEAW651446
Lyophilisationsampullen, Vacule®, WHEATON®
Ampullen
These lyophilisation ampoules are made of low extractable borosilicate glass.
  • Heat or stopper sealed
Ideal for small volume lyophilization samples.
Zertifizierungen: USP Type , ASTM E 438 Type I, Class A
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge