WHEATON® COMPLETEPACK Vials with Stoppers and Seals, Amber Glass
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
W991000-CPA W991001-CPA W991002-CPA W991003-CPA W991004-CPA W991005-CPA W991006-CPA W991007-CPA W991008-CPA W991009-CPA W991010-CPA W991011-CPA W991012-CPA W991013-CPA W991014-CPA W991015-CPA W991016-CPA W991017-CPA
WHEAW991000-CPAEA 4680 EUR InStock
WHEAW991000-CPA WHEAW991001-CPA WHEAW991002-CPA WHEAW991003-CPA WHEAW991004-CPA WHEAW991005-CPA WHEAW991006-CPA WHEAW991007-CPA WHEAW991008-CPA WHEAW991009-CPA WHEAW991010-CPA WHEAW991011-CPA WHEAW991012-CPA WHEAW991013-CPA WHEAW991014-CPA WHEAW991015-CPA WHEAW991016-CPA WHEAW991017-CPA
WHEATON® COMPLETEPACK Vials with Stoppers and Seals, Amber Glass
Vials
Complete range of amber crimp top vials, rubber stoppers and seals that are sterilised and ready-to-use.
  • Complete manufacturing and certificate of analysis traceability
  • Container Closure Integrity (CCI) validation
  • Amber glass vials especially for photosensitive contents
Ready-to-Use, Just-in-Time, minimum 180 day shelf life from time of shipment.
Zertifizierungen: USP <788>/EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP <85>/EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻⁶ (Vials, Stoppers, and Seals), USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge